Learn More
Meso Scale Discovery VPLEX SARSCoV2 PNL 1 ACE2 Kit
SDP
Catalog No. 5000081368
Encompass
The SARS-CoV-2 Panel 1 (ACE2) is a research use only multiplex immunoassay panel from Meso Scale Diagnostics that allows simultaneous measurement of biomarkers related to COVID-19 infection This 10-plex panel includes assays for IgA IgG and IgM antibodies to SARS-CoV-2 antigens as well as assays to measure levels of ACE2 and cytokines associated with COVID-19 disease severity The electrochemiluminescence immunoassays provide high sensitivity and broad dynamic range This panel could help researchers better understand immune responses and disease mechanisms in COVID-19
Catalog No. 50-000-81368 Supplier Meso Scale Discovery Supplier No. K15375U2
Add to Cart
Add to Cart
The SARS-CoV-2 Panel 1 (ACE2) is a research use only multiplex immunoassay panel from Meso Scale Diagnostics that allows simultaneous measurement of biomarkers related to COVID-19 infection This 10-plex panel includes assays for IgA IgG and IgM antibodies to SARS-CoV-2 antigens as well as assays to measure levels of ACE2 and cytokines associated with COVID-19 disease severity The electrochemiluminescence immunoassays provide high sensitivity and broad dynamic range This panel could help researchers better understand immune responses and disease mechanisms in COVID-19