Learn More
Meso Scale Discovery VPLEX SARSCoV2 PNL 1 IgA Kit
SDP
Catalog No. 5000081913
Encompass
The Meso Scale Discovery SARS-CoV-2 Panel 1 IgA is an electrochemiluminescence immunoassay kit for the qualitative detection of IgA antibodies to SARS-CoV-2 in human serum and plasma The assay uses Meso Scale Discovery s proprietary MULTI-ARRAY technology and provides a semiquantitative result reported as an index value relative to calibrator samples The panel allows for testing of up to 30 samples in duplicate along with calibrators and controls The assay has high sensitivity and specificity and takes approximately 4 hours to complete This product aids in identifying individuals with an adaptive immune response to SARS-CoV-2 indicating recent or prior infection
Catalog No. 50-000-81913 Supplier Meso Scale Discovery Supplier No. K15361U2
The Meso Scale Discovery SARS-CoV-2 Panel 1 IgA is an electrochemiluminescence immunoassay kit for the qualitative detection of IgA antibodies to SARS-CoV-2 in human serum and plasma The assay uses Meso Scale Discovery s proprietary MULTI-ARRAY technology and provides a semiquantitative result reported as an index value relative to calibrator samples The panel allows for testing of up to 30 samples in duplicate along with calibrators and controls The assay has high sensitivity and specificity and takes approximately 4 hours to complete This product aids in identifying individuals with an adaptive immune response to SARS-CoV-2 indicating recent or prior infection