Learn More
Meso Scale Discovery VPLEX SARSCoV2 PNL 1 IgG Kit
SDP
Catalog No. 5000079887
Encompass
The Meso Scale Discovery SARS-CoV-2 Panel 1 IgG is an immunoassay that detects IgG antibodies to SARS-CoV-2 in human serum and plasma This qualitative test uses MSD electrochemiluminescence technology and provides results in under 3 hours The panel contains a V-PLEX COVID-19 Coronavirus Panel 1 IgG Kit and a V-PLEX Human IgG Calibrator Kit It is intended for use as an aid in identifying individuals with an adaptive immune response to SARS-CoV-2 indicating recent or prior infection This product requires the MSD QuickPlex SQ120 Instrument
Catalog No. 50-000-79887 Supplier Meso Scale Discovery Supplier No. K15359U2
Add to Cart
Add to Cart
The Meso Scale Discovery SARS-CoV-2 Panel 1 IgG is an immunoassay that detects IgG antibodies to SARS-CoV-2 in human serum and plasma This qualitative test uses MSD electrochemiluminescence technology and provides results in under 3 hours The panel contains a V-PLEX COVID-19 Coronavirus Panel 1 IgG Kit and a V-PLEX Human IgG Calibrator Kit It is intended for use as an aid in identifying individuals with an adaptive immune response to SARS-CoV-2 indicating recent or prior infection This product requires the MSD QuickPlex SQ120 Instrument