Learn More
Meso Scale Discovery VPLEX SARSCoV2 PNL 1 IgG
SDP
Catalog No. 5000082049
Encompass
The Meso Scale Diagnostics SARS-CoV-2 Panel 1 IgG is an immunoassay test kit that detects IgG antibodies to SARS-CoV-2 in human serum and plasma It uses electrochemiluminescence technology on Meso Scale Diagnostics proprietary multi-array plate platform The test kit includes a 96-well plate coated with SARS-CoV-2 antigens detection antibodies calibrators controls diluent and wash buffer Results are obtained in under 3 hours using Meso Scale Diagnostics MESO QuickPlex SQ 120 instrument This semi-quantitative test can aid in identifying individuals with an adaptive immune response to SARS-CoV-2 indicating recent or prior infection
Catalog No. 50-000-82049 Supplier Meso Scale Discovery Supplier No. K15359U4
Add to Cart
Add to Cart
Request a Quote
The Meso Scale Diagnostics SARS-CoV-2 Panel 1 IgG is an immunoassay test kit that detects IgG antibodies to SARS-CoV-2 in human serum and plasma It uses electrochemiluminescence technology on Meso Scale Diagnostics proprietary multi-array plate platform The test kit includes a 96-well plate coated with SARS-CoV-2 antigens detection antibodies calibrators controls diluent and wash buffer Results are obtained in under 3 hours using Meso Scale Diagnostics MESO QuickPlex SQ 120 instrument This semi-quantitative test can aid in identifying individuals with an adaptive immune response to SARS-CoV-2 indicating recent or prior infection