Learn More
Meso Scale Discovery VPLEX SARSCoV2 PNL 1 IgM Kit
SDP
Catalog No. 5000081939
Encompass
The Meso Scale Diagnostics SARS-CoV-2 Panel 1 IgM is an in vitro diagnostic test for the qualitative detection of IgM antibodies to SARS-CoV-2 in human serum and plasma This electrochemiluminescence immunoassay utilizes magnetic beads coated with SARS-CoV-2 antigens to detect IgM antibodies that bind to the antigens The test is intended for use as an aid in identifying individuals with an adaptive immune response to SARS-CoV-2 indicating recent or prior infection The panel consists of a 96-well plate and all necessary reagents and can process 90 samples in approximately 3 hours This is a sensitive and specific assay that provides quantitative results
Catalog No. 50-000-81939 Supplier Meso Scale Discovery Supplier No. K15360U2
The Meso Scale Diagnostics SARS-CoV-2 Panel 1 IgM is an in vitro diagnostic test for the qualitative detection of IgM antibodies to SARS-CoV-2 in human serum and plasma This electrochemiluminescence immunoassay utilizes magnetic beads coated with SARS-CoV-2 antigens to detect IgM antibodies that bind to the antigens The test is intended for use as an aid in identifying individuals with an adaptive immune response to SARS-CoV-2 indicating recent or prior infection The panel consists of a 96-well plate and all necessary reagents and can process 90 samples in approximately 3 hours This is a sensitive and specific assay that provides quantitative results