Learn More
Meso Scale Discovery VPLEX SARSCoV2 PNL 1 IgM
SDP
Catalog No. 5000081505
Encompass
The Meso Scale Discovery SARS-CoV-2 Panel 1 IgM is an in vitro diagnostic test kit that detects IgM antibodies to SARS-CoV-2 in human serum and plasma This qualitative immunoassay uses electroluminescence technology on patterned arrays for detection of antibodies that bind to the receptor binding domain of the SARS-CoV-2 spike protein The test has high sensitivity and specificity and is intended to aid in identifying individuals with an adaptive immune response to SARS-CoV-2 indicating recent or prior infection The kit includes a 96-well plate coated with antigen detection antibody read buffer wash buffer calibrators and controls
Catalog No. 50-000-81505 Supplier Meso Scale Discovery Supplier No. K15360U4
Add to Cart
Add to Cart
Request a Quote
The Meso Scale Discovery SARS-CoV-2 Panel 1 IgM is an in vitro diagnostic test kit that detects IgM antibodies to SARS-CoV-2 in human serum and plasma This qualitative immunoassay uses electroluminescence technology on patterned arrays for detection of antibodies that bind to the receptor binding domain of the SARS-CoV-2 spike protein The test has high sensitivity and specificity and is intended to aid in identifying individuals with an adaptive immune response to SARS-CoV-2 indicating recent or prior infection The kit includes a 96-well plate coated with antigen detection antibody read buffer wash buffer calibrators and controls