Learn More
Meso Scale Discovery VPLEX SARSCoV2 PNL 1 MSE IgA
SDP
Catalog No. 5000078289
Encompass
The Meso Scale Discovery V-PLEX SARS-CoV-2 Panel 1 Mouse IgA Kit is an electrochemiluminescence immunoassay for quantifying IgA antibodies to SARS-CoV-2 in mouse serum and plasma It uses plates coated with spike protein S1 and nucleocapsid protein antigens to capture antibodies with detection via SULFO-TAG labeled anti-mouse IgA The kit includes reagents for 96 tests offering sensitivity comparable to ELISA Results are available in hours using an MSD instrument aiding vaccine and therapeutic development in mouse models
Catalog No. 50-000-78289 Supplier Meso Scale Discovery Supplier No. K15474U4
Add to Cart
Add to Cart
Request a Quote
The Meso Scale Discovery V-PLEX SARS-CoV-2 Panel 1 Mouse IgA Kit is an electrochemiluminescence immunoassay for quantifying IgA antibodies to SARS-CoV-2 in mouse serum and plasma It uses plates coated with spike protein S1 and nucleocapsid protein antigens to capture antibodies with detection via SULFO-TAG labeled anti-mouse IgA The kit includes reagents for 96 tests offering sensitivity comparable to ELISA Results are available in hours using an MSD instrument aiding vaccine and therapeutic development in mouse models