Learn More
Meso Scale Discovery VPLEX SARSCoV2 PNL 1 MSE IgG
SDP
Catalog No. 5000079772
Encompass
The Meso Scale Discovery V-PLEX SARS-CoV-2 Panel 1 Mouse IgG Kit is an electrochemiluminescence immunoassay that detects IgG antibodies to SARS-CoV-2 in mouse serum and plasma samples This assay uses the S1 and receptor binding domain (RBD) antigens to detect antibodies that bind to the spike protein The kit includes a 96-well plate coated with antigens detection antibody read buffer wash buffer calibrators and controls Results are obtained in 3 5 hours using an MSD instrument This assay allows high-throughput sensitive and quantitative measurement of SARS-CoV-2 IgG antibodies
Catalog No. 50-000-79772 Supplier Meso Scale Discovery Supplier No. K15472U4
Add to Cart
Add to Cart
Request a Quote
The Meso Scale Discovery V-PLEX SARS-CoV-2 Panel 1 Mouse IgG Kit is an electrochemiluminescence immunoassay that detects IgG antibodies to SARS-CoV-2 in mouse serum and plasma samples This assay uses the S1 and receptor binding domain (RBD) antigens to detect antibodies that bind to the spike protein The kit includes a 96-well plate coated with antigens detection antibody read buffer wash buffer calibrators and controls Results are obtained in 3 5 hours using an MSD instrument This assay allows high-throughput sensitive and quantitative measurement of SARS-CoV-2 IgG antibodies