Learn More
Meso Scale Discovery VPLEX SARSCoV2 PNL 1 MSE IgM
SDP
Catalog No. 5000080249
Encompass
The Meso Scale Discovery V-PLEX SARS-CoV-2 Panel 1 Mouse IgM Kit is an electrochemiluminescence immunoassay for detecting IgM antibodies to SARS-CoV-2 in mouse serum and plasma It uses a plate coated with SARS-CoV-2 spike protein to capture antibodies detected by an anti-mouse IgM antibody with an electrochemiluminescent label The kit includes reagents for 96 tests measuring IgM levels with high sensitivity and specificity The assay takes about 3 hours and requires a Meso Scale Discovery instrument This kit reliably detects SARS-CoV-2 specific IgM in mouse samples
Catalog No. 50-000-80249 Supplier Meso Scale Discovery Supplier No. K15473U4
Add to Cart
Add to Cart
Request a Quote
The Meso Scale Discovery V-PLEX SARS-CoV-2 Panel 1 Mouse IgM Kit is an electrochemiluminescence immunoassay for detecting IgM antibodies to SARS-CoV-2 in mouse serum and plasma It uses a plate coated with SARS-CoV-2 spike protein to capture antibodies detected by an anti-mouse IgM antibody with an electrochemiluminescent label The kit includes reagents for 96 tests measuring IgM levels with high sensitivity and specificity The assay takes about 3 hours and requires a Meso Scale Discovery instrument This kit reliably detects SARS-CoV-2 specific IgM in mouse samples