Learn More
Meso Scale Discovery VPLEX SARSCoV2 PNL 13 IgG
SDP
Catalog No. 5000081438
Encompass
The Mesoscale K15463U-4 is a 4-channel 16-bit 250 kS/s simultaneous sampling data acquisition module It has 4 analog voltage input channels with programmable gain amplifiers anti-aliasing filters and 16-bit ADCs The input voltage range is 10 V It communicates via USB and is compatible with Windows Linux and Mac operating systems The module is bus-powered compact and portable It is well suited for low-frequency data acquisition applications
Catalog No. 50-000-81438 Supplier Meso Scale Discovery Supplier No. K15463U4
Add to Cart
Add to Cart
Request a Quote
The Mesoscale K15463U-4 is a 4-channel 16-bit 250 kS/s simultaneous sampling data acquisition module It has 4 analog voltage input channels with programmable gain amplifiers anti-aliasing filters and 16-bit ADCs The input voltage range is 10 V It communicates via USB and is compatible with Windows Linux and Mac operating systems The module is bus-powered compact and portable It is well suited for low-frequency data acquisition applications