Learn More
Meso Scale Discovery VPLEX SARSCoV2 PNL 13 MSE IgA
SDP
Catalog No. 5000082318
Encompass
The Meso Scale Discovery V-PLEX SARS-CoV-2 Panel 13 Mouse IgA Kit is an electrochemiluminescence immunoassay that enables the simultaneous detection and quantification of IgA antibodies to 13 SARS-CoV-2 antigens in mouse serum plasma and other biological matrices This multiplex assay kit includes plates coated with SARS-CoV-2 spike protein S1 spike protein receptor binding domain (RBD) nucleocapsid protein and 10 additional SARS-CoV-2 antigens The kit provides a high-throughput sensitive and reproducible method to measure the IgA antibody response to SARS-CoV-2 infection or vaccination in mouse models
Catalog No. 50-000-82318 Supplier Meso Scale Discovery Supplier No. K15514U4
Add to Cart
Add to Cart
Request a Quote
The Meso Scale Discovery V-PLEX SARS-CoV-2 Panel 13 Mouse IgA Kit is an electrochemiluminescence immunoassay that enables the simultaneous detection and quantification of IgA antibodies to 13 SARS-CoV-2 antigens in mouse serum plasma and other biological matrices This multiplex assay kit includes plates coated with SARS-CoV-2 spike protein S1 spike protein receptor binding domain (RBD) nucleocapsid protein and 10 additional SARS-CoV-2 antigens The kit provides a high-throughput sensitive and reproducible method to measure the IgA antibody response to SARS-CoV-2 infection or vaccination in mouse models