Learn More
Meso Scale Discovery VPLEX SARSCoV2 PNL 13 MSE IgA
SDP
Catalog No. 5000082458
Encompass
The Mesoscale K15514U-2 is a high-performance thermoelectric cooler for photonics and semiconductor applications offering precise temperature control It has a 15W heat pumping capacity and a 70 C temperature differential With dimensions of 30 x 30 x 3 4mm it includes a thermistor for temperature sensing This Peltier cooler is ideal for stabilizing laser diodes and cooling sensors featuring a high coefficient of performance and low power consumption A heat sink and fan are required for heat dissipation from the hot side
Catalog No. 50-000-82458 Supplier Meso Scale Discovery Supplier No. K15514U2
Add to Cart
Add to Cart
Request a Quote
The Mesoscale K15514U-2 is a high-performance thermoelectric cooler for photonics and semiconductor applications offering precise temperature control It has a 15W heat pumping capacity and a 70 C temperature differential With dimensions of 30 x 30 x 3 4mm it includes a thermistor for temperature sensing This Peltier cooler is ideal for stabilizing laser diodes and cooling sensors featuring a high coefficient of performance and low power consumption A heat sink and fan are required for heat dissipation from the hot side