Learn More
Meso Scale Discovery VPLEX SARSCoV2 PNL 13 MSE IgG
SDP
Catalog No. 5000082317
Encompass
The Meso Scale Discovery V-PLEX SARS-CoV-2 Panel 13 Mouse IgG Kit is an electrochemiluminescence immunoassay that detects IgG antibodies to 13 SARS-CoV-2 antigens in mouse serum and plasma samples The kit includes a 96-well plate coated with SARS-CoV-2 spike protein S1 spike protein S2 receptor binding domain (RBD) and nucleocapsid protein antigens along with calibrators controls detection antibodies read buffer and other reagents required to run the assay Results are obtained in under 3 hours using Meso Scale Discovery instrumentation The kit allows for quantitative measurement of IgG antibodies elicited in response to SARS-CoV-2 infection or vaccination in mouse models
Catalog No. 50-000-82317 Supplier Meso Scale Discovery Supplier No. K15512U4
Add to Cart
Add to Cart
Request a Quote
The Meso Scale Discovery V-PLEX SARS-CoV-2 Panel 13 Mouse IgG Kit is an electrochemiluminescence immunoassay that detects IgG antibodies to 13 SARS-CoV-2 antigens in mouse serum and plasma samples The kit includes a 96-well plate coated with SARS-CoV-2 spike protein S1 spike protein S2 receptor binding domain (RBD) and nucleocapsid protein antigens along with calibrators controls detection antibodies read buffer and other reagents required to run the assay Results are obtained in under 3 hours using Meso Scale Discovery instrumentation The kit allows for quantitative measurement of IgG antibodies elicited in response to SARS-CoV-2 infection or vaccination in mouse models