Learn More
Meso Scale Discovery VPLEX SARSCoV2 PNL 13 MSE IgM
SDP
Catalog No. 5000081538
Encompass
The Mesoscale K15513U-4 is a 4-channel 16-bit 250 kS/s simultaneous sampling data acquisition module It has 4 analog voltage input channels with programmable input ranges of 10 V 5 V 2 V and 1 V The module has 16-bit resolution and a maximum sampling rate of 250 kS/s It can be used for applications such as sensor measurements analog signal monitoring and data logging The module has USB connectivity for easy integration with PCs Key features include 4 simultaneous sampling channels 16-bit ADC resolution 250 kS/s max sampling rate and USB interface
Catalog No. 50-000-81538 Supplier Meso Scale Discovery Supplier No. K15513U4
Add to Cart
Add to Cart
Request a Quote
The Mesoscale K15513U-4 is a 4-channel 16-bit 250 kS/s simultaneous sampling data acquisition module It has 4 analog voltage input channels with programmable input ranges of 10 V 5 V 2 V and 1 V The module has 16-bit resolution and a maximum sampling rate of 250 kS/s It can be used for applications such as sensor measurements analog signal monitoring and data logging The module has USB connectivity for easy integration with PCs Key features include 4 simultaneous sampling channels 16-bit ADC resolution 250 kS/s max sampling rate and USB interface