Learn More
Meso Scale Discovery VPLEX SARSCoV2 PNL 2 ACE2 Kit
SDP
Catalog No. 5000082044
Encompass
The SARS-CoV-2 Panel 2 (ACE2) is a multiplex immunoassay panel from Meso Scale Diagnostics that detects antibodies to the SARS-CoV-2 spike and nucleocapsid proteins as well as the ACE2 receptor It uses electrochemiluminescence detection technology and provides quantitative results from human serum plasma or whole blood samples The panel includes reagents for 96 tests and can help evaluate immune responses in COVID-19 vaccine studies and natural infection It has high sensitivity and specificity and has received Emergency Use Authorization from the FDA
Catalog No. 50-000-82044 Supplier Meso Scale Discovery Supplier No. K15386U2
The SARS-CoV-2 Panel 2 (ACE2) is a multiplex immunoassay panel from Meso Scale Diagnostics that detects antibodies to the SARS-CoV-2 spike and nucleocapsid proteins as well as the ACE2 receptor It uses electrochemiluminescence detection technology and provides quantitative results from human serum plasma or whole blood samples The panel includes reagents for 96 tests and can help evaluate immune responses in COVID-19 vaccine studies and natural infection It has high sensitivity and specificity and has received Emergency Use Authorization from the FDA