Learn More
Meso Scale Discovery VPLEX SARSCoV2 PNL 2 ACE2
SDP
Catalog No. 5000079804
Encompass
The SARS-CoV-2 Panel 2 (ACE2) by Meso Scale Diagnostics is a multiplex immunoassay for detecting IgG antibodies to SARS-CoV-2 antigens in human serum and plasma It includes a V-PLEX COVID-19 Coronavirus Panel 2 (IgG) kit with an anti-human IgG detection antibody recombinant SARS-CoV-2 RBD antigen nucleocapsid protein antigen and quality controls This panel enables qualitative and semi-quantitative detection of IgG antibodies specific to SARS-CoV-2 RBD and nucleocapsid protein aiding in identifying individuals with an adaptive immune response to SARS-CoV-2
Catalog No. 50-000-79804 Supplier Meso Scale Discovery Supplier No. K15386U4
Add to Cart
Add to Cart
Request a Quote
The SARS-CoV-2 Panel 2 (ACE2) by Meso Scale Diagnostics is a multiplex immunoassay for detecting IgG antibodies to SARS-CoV-2 antigens in human serum and plasma It includes a V-PLEX COVID-19 Coronavirus Panel 2 (IgG) kit with an anti-human IgG detection antibody recombinant SARS-CoV-2 RBD antigen nucleocapsid protein antigen and quality controls This panel enables qualitative and semi-quantitative detection of IgG antibodies specific to SARS-CoV-2 RBD and nucleocapsid protein aiding in identifying individuals with an adaptive immune response to SARS-CoV-2