Learn More
Meso Scale Discovery VPLEX SARSCoV2 PNL 2 IgA Kit
SDP
Catalog No. 5000082186
Encompass
The Meso Scale Discovery SARS-CoV-2 Panel 2 IgA is an in vitro diagnostic test kit that detects IgA antibodies to SARS-CoV-2 in human serum and plasma It uses electrochemiluminescence technology and provides quantitative results for IgA antibodies to the receptor binding domain and nucleocapsid protein of SARS-CoV-2 The test kit includes a 96-well plate coated with antigens detection antibody read buffer wash buffer calibrators and controls It can aid in identifying individuals with an adaptive immune response to SARS-CoV-2 indicating recent or prior infection The test has high sensitivity and specificity and is for use under an Emergency Use Authorization only
Catalog No. 50-000-82186 Supplier Meso Scale Discovery Supplier No. K15385U2
The Meso Scale Discovery SARS-CoV-2 Panel 2 IgA is an in vitro diagnostic test kit that detects IgA antibodies to SARS-CoV-2 in human serum and plasma It uses electrochemiluminescence technology and provides quantitative results for IgA antibodies to the receptor binding domain and nucleocapsid protein of SARS-CoV-2 The test kit includes a 96-well plate coated with antigens detection antibody read buffer wash buffer calibrators and controls It can aid in identifying individuals with an adaptive immune response to SARS-CoV-2 indicating recent or prior infection The test has high sensitivity and specificity and is for use under an Emergency Use Authorization only