Learn More
Meso Scale Discovery VPLEX SARSCoV2 PNL 2 IgA
SDP
Catalog No. 5000081969
Encompass
The Meso Scale Diagnostics SARS-CoV-2 Panel 2 IgA is an in vitro diagnostic test for the qualitative detection of IgA antibodies to SARS-CoV-2 in human serum and plasma This electrochemiluminescence immunoassay utilizes recombinant SARS-CoV-2 antigens to detect IgA antibodies that bind to the receptor binding domain and nucleocapsid protein The test is intended for use as an aid in identifying individuals with an adaptive immune response to SARS-CoV-2 indicating recent or prior infection Results should be used in conjunction with clinical findings and other laboratory tests This product requires the Meso Scale Discovery MULTI-ARRAY 96-well Plate and MSD Reader
Catalog No. 50-000-81969 Supplier Meso Scale Discovery Supplier No. K15385U4
Add to Cart
Add to Cart
Request a Quote
The Meso Scale Diagnostics SARS-CoV-2 Panel 2 IgA is an in vitro diagnostic test for the qualitative detection of IgA antibodies to SARS-CoV-2 in human serum and plasma This electrochemiluminescence immunoassay utilizes recombinant SARS-CoV-2 antigens to detect IgA antibodies that bind to the receptor binding domain and nucleocapsid protein The test is intended for use as an aid in identifying individuals with an adaptive immune response to SARS-CoV-2 indicating recent or prior infection Results should be used in conjunction with clinical findings and other laboratory tests This product requires the Meso Scale Discovery MULTI-ARRAY 96-well Plate and MSD Reader