Learn More
Meso Scale Discovery VPLEX SARSCoV2 PNL 2 IgG
SDP
Catalog No. 5000082080
Encompass
The Meso Scale Discovery SARS-CoV-2 Panel 2 IgG is an in vitro diagnostic test kit that detects IgG antibodies to SARS-CoV-2 in human serum and plasma It uses electrochemiluminescence technology and provides quantitative results for IgG antibodies to the spike and nucleocapsid proteins The test has high sensitivity and specificity and is intended to aid in identifying individuals with an adaptive immune response to SARS-CoV-2 indicating recent or prior infection Results are available in 2 5 hours using an MSD reader This product requires refrigerated storage and is for use under an Emergency Use Authorization only
Catalog No. 50-000-82080 Supplier Meso Scale Discovery Supplier No. K15383U4
Add to Cart
Add to Cart
Request a Quote
The Meso Scale Discovery SARS-CoV-2 Panel 2 IgG is an in vitro diagnostic test kit that detects IgG antibodies to SARS-CoV-2 in human serum and plasma It uses electrochemiluminescence technology and provides quantitative results for IgG antibodies to the spike and nucleocapsid proteins The test has high sensitivity and specificity and is intended to aid in identifying individuals with an adaptive immune response to SARS-CoV-2 indicating recent or prior infection Results are available in 2 5 hours using an MSD reader This product requires refrigerated storage and is for use under an Emergency Use Authorization only