Learn More
Meso Scale Discovery VPLEX SARSCoV2 PNL 2 IgM Kit
SDP
Catalog No. 5000081504
Encompass
The Meso Scale Diagnostics SARS-CoV-2 Panel 2 IgM is an in vitro diagnostic test for detecting IgM antibodies to SARS-CoV-2 in human serum and plasma This electrochemiluminescence immunoassay uses magnetic beads with SARS-CoV-2 antigens to capture IgM antibodies for spike and nucleocapsid proteins Results are ready in 2 5 hours with an MSD reader It shows 99 1% positive and 99 4% negative agreement versus PCR The test requires the MSD Discovery Workbench and an MSD reader
Catalog No. 50-000-81504 Supplier Meso Scale Discovery Supplier No. K15384U2
The Meso Scale Diagnostics SARS-CoV-2 Panel 2 IgM is an in vitro diagnostic test for detecting IgM antibodies to SARS-CoV-2 in human serum and plasma This electrochemiluminescence immunoassay uses magnetic beads with SARS-CoV-2 antigens to capture IgM antibodies for spike and nucleocapsid proteins Results are ready in 2 5 hours with an MSD reader It shows 99 1% positive and 99 4% negative agreement versus PCR The test requires the MSD Discovery Workbench and an MSD reader