Learn More
Meso Scale Discovery VPLEX SARSCoV2 PNL 2 IgM
SDP
Catalog No. 5000078589
Encompass
The Meso Scale Diagnostics SARS-CoV-2 Panel 2 IgM is an in vitro diagnostic test for the qualitative detection of IgM antibodies to SARS-CoV-2 in human serum and plasma This electrochemiluminescence immunoassay utilizes a recombinant SARS-CoV-2 nucleocapsid antigen to detect IgM antibodies that are generated by the immune system in response to a SARS-CoV-2 infection The test is intended for use as an aid in identifying individuals with an adaptive immune response to SARS-CoV-2 indicating recent or prior infection It has not been FDA cleared or approved and has been authorized by FDA under an EUA for use by authorized laboratories
Catalog No. 50-000-78589 Supplier Meso Scale Discovery Supplier No. K15384U4
Add to Cart
Add to Cart
Request a Quote
The Meso Scale Diagnostics SARS-CoV-2 Panel 2 IgM is an in vitro diagnostic test for the qualitative detection of IgM antibodies to SARS-CoV-2 in human serum and plasma This electrochemiluminescence immunoassay utilizes a recombinant SARS-CoV-2 nucleocapsid antigen to detect IgM antibodies that are generated by the immune system in response to a SARS-CoV-2 infection The test is intended for use as an aid in identifying individuals with an adaptive immune response to SARS-CoV-2 indicating recent or prior infection It has not been FDA cleared or approved and has been authorized by FDA under an EUA for use by authorized laboratories