Learn More
Meso Scale Discovery VPLEX SARSCoV2 PNL 2 MSE IgA
SDP
Catalog No. 5000081566
Encompass
The V-PLEX Human Cytokine 30-Plex Kit is an immunoassay that allows simultaneous measurement of 30 human cytokines and chemokines in serum plasma and culture supernatants It uses electrochemiluminescence detection technology and provides high sensitivity broad dynamic range and requires low sample volume The kit includes a 96-well plate coated with capture antibodies detection antibodies labeled with ruthenium and calibrator and control solutions It can quantify cytokines like IL-1 IL-2 IL-4 IL-5 IL-6 IL-7 IL-8 IL-10 IL-12p70 IL-13 IL-17A TNF- IFN- GM-CSF and more The kit has a short 5-hour assay time from sample to results
Catalog No. 50-000-81566 Supplier Meso Scale Discovery Supplier No. K15477U2
The V-PLEX Human Cytokine 30-Plex Kit is an immunoassay that allows simultaneous measurement of 30 human cytokines and chemokines in serum plasma and culture supernatants It uses electrochemiluminescence detection technology and provides high sensitivity broad dynamic range and requires low sample volume The kit includes a 96-well plate coated with capture antibodies detection antibodies labeled with ruthenium and calibrator and control solutions It can quantify cytokines like IL-1 IL-2 IL-4 IL-5 IL-6 IL-7 IL-8 IL-10 IL-12p70 IL-13 IL-17A TNF- IFN- GM-CSF and more The kit has a short 5-hour assay time from sample to results