Learn More
Meso Scale Discovery VPLEX SARSCoV2 PNL 2 MSE IgG
SDP
Catalog No. 5000079769
Encompass
The Meso Scale Discovery V-PLEX SARS-CoV-2 Panel 2 Mouse IgG Kit is an electrochemiluminescence immunoassay that detects IgG antibodies to SARS-CoV-2 in mouse serum and plasma samples This assay uses a multiplex format to measure antibodies specific for the SARS-CoV-2 spike protein receptor binding domain and nucleocapsid protein The kit includes a plate coated with antigens detection antibodies calibrators controls diluent read buffer and wash buffer Results are obtained in under 3 hours using an MSD instrument This assay allows for high-throughput sensitive and reproducible measurement of SARS-CoV-2 IgG antibodies in mouse models
Catalog No. 50-000-79769 Supplier Meso Scale Discovery Supplier No. K15475U4
Add to Cart
Add to Cart
Request a Quote
The Meso Scale Discovery V-PLEX SARS-CoV-2 Panel 2 Mouse IgG Kit is an electrochemiluminescence immunoassay that detects IgG antibodies to SARS-CoV-2 in mouse serum and plasma samples This assay uses a multiplex format to measure antibodies specific for the SARS-CoV-2 spike protein receptor binding domain and nucleocapsid protein The kit includes a plate coated with antigens detection antibodies calibrators controls diluent read buffer and wash buffer Results are obtained in under 3 hours using an MSD instrument This assay allows for high-throughput sensitive and reproducible measurement of SARS-CoV-2 IgG antibodies in mouse models