Learn More
Meso Scale Discovery VPLEX SARSCoV2 PNL 2 MSE IgG
SDP
Catalog No. 5000080246
Encompass
The Meso Scale Discovery V-PLEX SARS-CoV-2 Panel 2 Mouse IgG Kit is an electrochemiluminescence immunoassay that detects IgG antibodies to SARS-CoV-2 in mouse serum and plasma samples The assay uses a multiplex format to measure antibodies specific for the SARS-CoV-2 spike protein receptor binding domain and nucleocapsid protein The kit includes a plate coated with antigens detection antibodies calibrators controls buffers and substrate The assay has a testing time of approximately 3 5 hours and provides quantitative results for SARS-CoV-2 IgG levels This product enables sensitive and specific measurement of the mouse antibody response to SARS-CoV-2 immunization or infection
Catalog No. 50-000-80246 Supplier Meso Scale Discovery Supplier No. K15475U2
The Meso Scale Discovery V-PLEX SARS-CoV-2 Panel 2 Mouse IgG Kit is an electrochemiluminescence immunoassay that detects IgG antibodies to SARS-CoV-2 in mouse serum and plasma samples The assay uses a multiplex format to measure antibodies specific for the SARS-CoV-2 spike protein receptor binding domain and nucleocapsid protein The kit includes a plate coated with antigens detection antibodies calibrators controls buffers and substrate The assay has a testing time of approximately 3 5 hours and provides quantitative results for SARS-CoV-2 IgG levels This product enables sensitive and specific measurement of the mouse antibody response to SARS-CoV-2 immunization or infection