Learn More
Meso Scale Discovery VPLEX SARSCoV2 PNL 2 MSE IgM
SDP
Catalog No. 5000080247
Encompass
The Meso Scale Discovery V-PLEX SARS-CoV-2 Panel 2 Mouse IgM Kit is an electrochemiluminescence immunoassay that detects IgM antibodies to SARS-CoV-2 in mouse serum and plasma samples The assay uses a plate coated with SARS-CoV-2 spike protein antigens to capture anti-SARS-CoV-2 antibodies from samples Detection is done using an anti-mouse IgM detection antibody conjugated to an electrochemiluminescent label The kit includes reagents for 96 tests and can quantitatively measure IgM levels to SARS-CoV-2 with high sensitivity and specificity The assay takes approximately 3 hours to complete
Catalog No. 50-000-80247 Supplier Meso Scale Discovery Supplier No. K15476U2
The Meso Scale Discovery V-PLEX SARS-CoV-2 Panel 2 Mouse IgM Kit is an electrochemiluminescence immunoassay that detects IgM antibodies to SARS-CoV-2 in mouse serum and plasma samples The assay uses a plate coated with SARS-CoV-2 spike protein antigens to capture anti-SARS-CoV-2 antibodies from samples Detection is done using an anti-mouse IgM detection antibody conjugated to an electrochemiluminescent label The kit includes reagents for 96 tests and can quantitatively measure IgM levels to SARS-CoV-2 with high sensitivity and specificity The assay takes approximately 3 hours to complete