Learn More
Meso Scale Discovery VPLEX SARSCoV2 PNL 2 MSE IgM
SDP
Catalog No. 5000081412
Encompass
The V-PLEX SARS-CoV-2 Panel 2 Mouse IgM Kit from Meso Scale Diagnostics is an electrochemiluminescence immunoassay that detects IgM antibodies to SARS-CoV-2 in mouse serum and plasma samples The assay uses a recombinant SARS-CoV-2 nucleocapsid protein to detect antibodies that bind to the nucleocapsid protein The kit includes a 96-well plate coated with the antigen detection antibody calibrators controls diluent read buffer wash buffer and plate seals The assay has a testing time of approximately 3 hours and 10 minutes and a sample dilution of 1 400 The kit can aid in evaluating vaccine efficacy and immunogenicity in mouse models of COVID-19
Catalog No. 50-000-81412 Supplier Meso Scale Discovery Supplier No. K15476U4
Add to Cart
Add to Cart
Request a Quote
The V-PLEX SARS-CoV-2 Panel 2 Mouse IgM Kit from Meso Scale Diagnostics is an electrochemiluminescence immunoassay that detects IgM antibodies to SARS-CoV-2 in mouse serum and plasma samples The assay uses a recombinant SARS-CoV-2 nucleocapsid protein to detect antibodies that bind to the nucleocapsid protein The kit includes a 96-well plate coated with the antigen detection antibody calibrators controls diluent read buffer wash buffer and plate seals The assay has a testing time of approximately 3 hours and 10 minutes and a sample dilution of 1 400 The kit can aid in evaluating vaccine efficacy and immunogenicity in mouse models of COVID-19