Learn More
Meso Scale Discovery VPLEX SARSCoV2 PNL 22 IgA Kit
SDP
Catalog No. 5000079765
Encompass
The Mesoscale K15561U-2 is a high-performance thermoelectric cooler for photonics and semiconductor applications offering precise temperature control It has a heat pumping capacity of 15 5W and a temperature differential of 70 C With dimensions of 30 x 30 x 3 4mm it requires 2 7VDC and handles up to 6A Operating between -20 C and 80 C it features low thermal resistance high reliability and RoHS compliance This Peltier cooler is ideal for stabilizing laser diodes and cooling sensors in temperature-critical applications
Catalog No. 50-000-79765 Supplier Meso Scale Discovery Supplier No. K15561U2
The Mesoscale K15561U-2 is a high-performance thermoelectric cooler for photonics and semiconductor applications offering precise temperature control It has a heat pumping capacity of 15 5W and a temperature differential of 70 C With dimensions of 30 x 30 x 3 4mm it requires 2 7VDC and handles up to 6A Operating between -20 C and 80 C it features low thermal resistance high reliability and RoHS compliance This Peltier cooler is ideal for stabilizing laser diodes and cooling sensors in temperature-critical applications