Learn More
Meso Scale Discovery VPLEX SARSCoV2 PNL 22 IgG Kit
SDP
Supplier:  Meso Scale Discovery K15559U2
Encompass
The Meso Scale Discovery V-PLEX Human MDC Kit is an electrochemiluminescent immunoassay for measuring Macrophage-Derived Chemokine (MDC) in human serum plasma and cell culture supernatants It includes a 96-well plate with capture antibody detection antibody with an electrochemiluminescent label calibrators diluent wash buffer read buffer and plate seals The assay s sensitivity is 0 3 pg/mL with an upper quantification limit of 10 000 pg/mL It uses MSD s MULTI-ARRAY technology offering high sensitivity wide dynamic range and low sample volume needs with high specificity for human MDC
Catalog No. 50-000-82373