Learn More
Meso Scale Discovery VPLEX SARSCoV2 PNL 22 MSE IgA
SDP
Catalog No. 5000079770
Encompass
The Meso Scale Discovery V-PLEX SARS-CoV-2 Panel 2 (2G) Mouse IgA Kit is an electrochemiluminescence immunoassay that detects IgA antibodies to SARS-CoV-2 in mouse serum and plasma samples The assay uses plates coated with SARS-CoV-2 spike protein S1 and nucleocapsid protein antigens to capture anti-SARS-CoV-2 antibodies in the sample Bound antibodies are detected using an anti-mouse IgA detection antibody conjugated to an electrochemiluminescent label The kit includes reagents for 25 tests and can quantitatively measure IgA antibodies with high sensitivity and specificity
Catalog No. 50-000-79770 Supplier Meso Scale Discovery Supplier No. K15565U4
Add to Cart
Add to Cart
Request a Quote
The Meso Scale Discovery V-PLEX SARS-CoV-2 Panel 2 (2G) Mouse IgA Kit is an electrochemiluminescence immunoassay that detects IgA antibodies to SARS-CoV-2 in mouse serum and plasma samples The assay uses plates coated with SARS-CoV-2 spike protein S1 and nucleocapsid protein antigens to capture anti-SARS-CoV-2 antibodies in the sample Bound antibodies are detected using an anti-mouse IgA detection antibody conjugated to an electrochemiluminescent label The kit includes reagents for 25 tests and can quantitatively measure IgA antibodies with high sensitivity and specificity