Learn More
Meso Scale Discovery VPLEX SARSCoV2 PNL 22 MSE IgA
SDP
Catalog No. 5000081458
Encompass
The Meso Scale Discovery COVID-19 Spike Protein Neutralization Assay Kit is designed to detect neutralizing antibodies in human serum plasma and other biological fluids The assay uses recombinant SARS-CoV-2 spike protein coated onto MSD GOLD electrodes to capture antibodies capable of blocking the ACE2 host cell receptor binding domain on the spike protein The assay has high sensitivity and specificity for detecting neutralizing antibodies and takes just a few hours to complete without the need for live virus The kit includes a 96-well plate coated with spike protein detection antibody read buffer sample diluent controls and calibrators
Catalog No. 50-000-81458 Supplier Meso Scale Discovery Supplier No. K15565U2
The Meso Scale Discovery COVID-19 Spike Protein Neutralization Assay Kit is designed to detect neutralizing antibodies in human serum plasma and other biological fluids The assay uses recombinant SARS-CoV-2 spike protein coated onto MSD GOLD electrodes to capture antibodies capable of blocking the ACE2 host cell receptor binding domain on the spike protein The assay has high sensitivity and specificity for detecting neutralizing antibodies and takes just a few hours to complete without the need for live virus The kit includes a 96-well plate coated with spike protein detection antibody read buffer sample diluent controls and calibrators