Learn More
Meso Scale Discovery VPLEX SARSCoV2 PNL 22 MSE IgM
SDP
Catalog No. 5000081457
Encompass
The Mesoscale K15564U-4 is a 4-channel 16-bit 250 kS/s simultaneous sampling data acquisition system It has 4 analog input channels with software-selectable input ranges of 10 V 5 V 2 V and 1 V The system can stream acquired data to PC memory at rates up to 250 kS/s per channel It connects to the host computer via USB 2 0 interface and is powered via this USB connection The K15564U-4 is compatible with Windows operating systems Key features include 16-bit A/D conversion software-selectable input ranges simultaneous sampling on all channels and streaming acquired data to PC memory The compact device is bus-powered for portability
Catalog No. 50-000-81457 Supplier Meso Scale Discovery Supplier No. K15564U4
Add to Cart
Add to Cart
Request a Quote
The Mesoscale K15564U-4 is a 4-channel 16-bit 250 kS/s simultaneous sampling data acquisition system It has 4 analog input channels with software-selectable input ranges of 10 V 5 V 2 V and 1 V The system can stream acquired data to PC memory at rates up to 250 kS/s per channel It connects to the host computer via USB 2 0 interface and is powered via this USB connection The K15564U-4 is compatible with Windows operating systems Key features include 16-bit A/D conversion software-selectable input ranges simultaneous sampling on all channels and streaming acquired data to PC memory The compact device is bus-powered for portability