Learn More
Meso Scale Discovery VPLEX SARSCoV2 PNL 23 ACE2 Kit
SDP
Catalog No. 5000082455
Encompass
The Mesoscale K15570U-2 is a high-performance thermoelectric cooler for photonics and semiconductor applications offering precise temperature control It has a heat pumping capacity of 15 5W and a temperature differential of 70 C With dimensions of 30 x 30 x 3 4mm it requires 2 7V and handles up to 6A The K15570U-2 uses a Bi2Te3 thermoelectric module and aluminum nitride ceramic substrate for optimal performance It is ideal for stabilizing laser diodes and maintaining CCD sensors at precise temperatures
Catalog No. 50-000-82455 Supplier Meso Scale Discovery Supplier No. K15570U2
Add to Cart
Add to Cart
Request a Quote
The Mesoscale K15570U-2 is a high-performance thermoelectric cooler for photonics and semiconductor applications offering precise temperature control It has a heat pumping capacity of 15 5W and a temperature differential of 70 C With dimensions of 30 x 30 x 3 4mm it requires 2 7V and handles up to 6A The K15570U-2 uses a Bi2Te3 thermoelectric module and aluminum nitride ceramic substrate for optimal performance It is ideal for stabilizing laser diodes and maintaining CCD sensors at precise temperatures