Learn More
Meso Scale Discovery VPLEX SARSCoV2 PNL 23 IgA
SDP
Catalog No. 5000080243
Encompass
The Meso Scale Diagnostics V-PLEX SARS-CoV-2 Panel 2 (IgA) Kit is an electrochemiluminescence immunoassay that detects IgA antibodies to SARS-CoV-2 in human serum and plasma This qualitative assay can aid in identifying individuals with an adaptive immune response to SARS-CoV-2 indicating recent or prior infection The kit includes a 4-spot MULTI-ARRAY 96-well plate coated with SARS-CoV-2 antigen detection antibodies calibrators and controls Results are analyzed using MSD s imaging systems and software This research use only assay provides a sensitive and rapid way to measure IgA antibodies
Catalog No. 50-000-80243 Supplier Meso Scale Discovery Supplier No. K15569U4
Add to Cart
Add to Cart
Request a Quote
The Meso Scale Diagnostics V-PLEX SARS-CoV-2 Panel 2 (IgA) Kit is an electrochemiluminescence immunoassay that detects IgA antibodies to SARS-CoV-2 in human serum and plasma This qualitative assay can aid in identifying individuals with an adaptive immune response to SARS-CoV-2 indicating recent or prior infection The kit includes a 4-spot MULTI-ARRAY 96-well plate coated with SARS-CoV-2 antigen detection antibodies calibrators and controls Results are analyzed using MSD s imaging systems and software This research use only assay provides a sensitive and rapid way to measure IgA antibodies