Learn More
Meso Scale Discovery VPLEX SARSCoV2 PNL 23 IgG Kit
SDP
Catalog No. 5000082319
Encompass
The Simoa SARS-CoV-2 N Protein Assay is a highly sensitive immunoassay that detects the nucleocapsid protein of SARS-CoV-2 in serum plasma whole blood and saliva It uses Quanterix s Simoa technology which is an ultrasensitive digital immunoassay that can detect proteins at femtomolar concentrations The assay has a limit of detection of 0 01 pg/mL and a linear range of 0 01-1000 pg/mL It has near 100% clinical sensitivity and specificity The assay can aid in diagnosing COVID-19 infection and monitoring disease progression and treatment response It provides quantitative results in around 3 hours This assay will be useful for sensitive detection of SARS-CoV-2 infection
Catalog No. 50-000-82319 Supplier Meso Scale Discovery Supplier No. K15567U2
Add to Cart
Add to Cart
Request a Quote
The Simoa SARS-CoV-2 N Protein Assay is a highly sensitive immunoassay that detects the nucleocapsid protein of SARS-CoV-2 in serum plasma whole blood and saliva It uses Quanterix s Simoa technology which is an ultrasensitive digital immunoassay that can detect proteins at femtomolar concentrations The assay has a limit of detection of 0 01 pg/mL and a linear range of 0 01-1000 pg/mL It has near 100% clinical sensitivity and specificity The assay can aid in diagnosing COVID-19 infection and monitoring disease progression and treatment response It provides quantitative results in around 3 hours This assay will be useful for sensitive detection of SARS-CoV-2 infection