Learn More
Meso Scale Discovery VPLEX SARSCoV2 PNL 23 IgG
SDP
Catalog No. 5000081567
Encompass
The Meso Scale Discovery COVID-19 Spike Protein Antibody Test Kit is an electrochemiluminescence immunoassay for detecting antibodies to the SARS-CoV-2 spike protein in human serum plasma and whole blood It uses MULTI-ARRAY technology providing quantitative results in under 3 hours The test is highly sensitive and specific for IgG antibodies aiding in identifying individuals with an adaptive immune response to SARS-CoV-2 The kit includes a 96-well plate with spike protein antigen detection antibodies calibrators controls and necessary reagents
Catalog No. 50-000-81567 Supplier Meso Scale Discovery Supplier No. K15567U4
Add to Cart
Add to Cart
Request a Quote
The Meso Scale Discovery COVID-19 Spike Protein Antibody Test Kit is an electrochemiluminescence immunoassay for detecting antibodies to the SARS-CoV-2 spike protein in human serum plasma and whole blood It uses MULTI-ARRAY technology providing quantitative results in under 3 hours The test is highly sensitive and specific for IgG antibodies aiding in identifying individuals with an adaptive immune response to SARS-CoV-2 The kit includes a 96-well plate with spike protein antigen detection antibodies calibrators controls and necessary reagents