Learn More
Meso Scale Discovery VPLEX SARSCoV2 PNL 23 IgM Kit
SDP
Catalog No. 5000079771
Encompass
The Meso Scale Discovery V-PLEX SARS-CoV-2 Panel 2 (IgM) is an electrochemiluminescent immunoassay kit that enables the quantitative measurement of IgM antibodies to SARS-CoV-2 in human serum and plasma This 5-plate kit includes reagents for 96 tests and can detect IgM antibodies targeting the SARS-CoV-2 nucleocapsid protein and spike protein receptor binding domain The assay has a testing time of approximately 3 5 hours and demonstrates sensitivity and specificity greater than 95% 14 days post symptom onset when compared to PCR This product aims to aid in understanding immune response to SARS-CoV-2 infection
Catalog No. 50-000-79771 Supplier Meso Scale Discovery Supplier No. K15568U2
Add to Cart
Add to Cart
Request a Quote
The Meso Scale Discovery V-PLEX SARS-CoV-2 Panel 2 (IgM) is an electrochemiluminescent immunoassay kit that enables the quantitative measurement of IgM antibodies to SARS-CoV-2 in human serum and plasma This 5-plate kit includes reagents for 96 tests and can detect IgM antibodies targeting the SARS-CoV-2 nucleocapsid protein and spike protein receptor binding domain The assay has a testing time of approximately 3 5 hours and demonstrates sensitivity and specificity greater than 95% 14 days post symptom onset when compared to PCR This product aims to aid in understanding immune response to SARS-CoV-2 infection