Learn More
Meso Scale Discovery VPLEX SARSCoV2 PNL 23 IgM
SDP
Catalog No. 5000078235
Encompass
The Meso Scale Discovery V-PLEX SARS-CoV-2 Panel 2 (IgM) is an electrochemiluminescent immunoassay kit that detects IgM antibodies to SARS-CoV-2 in human serum and plasma It includes a 96-well plate coated with SARS-CoV-2 antigens detection antibodies calibrators controls diluent wash buffer read buffer and plate seals The assay has a testing time of approximately 3 hours and 30 minutes and a sample volume requirement of 50 L It has a reportable range of 50-100 000 U/mL The kit includes sufficient reagents for one 96-well plate
Catalog No. 50-000-78235 Supplier Meso Scale Discovery Supplier No. K15568U4
Add to Cart
Add to Cart
Request a Quote
The Meso Scale Discovery V-PLEX SARS-CoV-2 Panel 2 (IgM) is an electrochemiluminescent immunoassay kit that detects IgM antibodies to SARS-CoV-2 in human serum and plasma It includes a 96-well plate coated with SARS-CoV-2 antigens detection antibodies calibrators controls diluent wash buffer read buffer and plate seals The assay has a testing time of approximately 3 hours and 30 minutes and a sample volume requirement of 50 L It has a reportable range of 50-100 000 U/mL The kit includes sufficient reagents for one 96-well plate