Learn More
Meso Scale Discovery VPLEX SARSCoV2 PNL 23 MSE IgA
SDP
Catalog No. 5000081414
Encompass
The V-PLEX Mouse Cytokine 30-Plex Kit from Meso Scale Discovery is a multiplex immunoassay for measuring 30 mouse cytokines and chemokines in one well It uses electrochemiluminescence detection for high sensitivity and broad dynamic range with low sample volume The kit includes a 96-well plate with capture antibodies detection antibodies labeled with an electrochemiluminescent compound and calibrator solutions for 30 analytes like IL-1 IL-2 IL-6 IFN- TNF- and more providing quantitative results from a small sample
Catalog No. 50-000-81414 Supplier Meso Scale Discovery Supplier No. K15573U4
Add to Cart
Add to Cart
Request a Quote
The V-PLEX Mouse Cytokine 30-Plex Kit from Meso Scale Discovery is a multiplex immunoassay for measuring 30 mouse cytokines and chemokines in one well It uses electrochemiluminescence detection for high sensitivity and broad dynamic range with low sample volume The kit includes a 96-well plate with capture antibodies detection antibodies labeled with an electrochemiluminescent compound and calibrator solutions for 30 analytes like IL-1 IL-2 IL-6 IFN- TNF- and more providing quantitative results from a small sample