Learn More
Meso Scale Discovery VPLEX SARSCoV2 PNL 23 MSE IgM
SDP
Catalog No. 5000081164
Encompass
The Meso Scale Discovery V-PLEX SARS-CoV-2 Panel 2 Mouse IgA Kit is a multiplex immunoassay that measures IgA antibodies to SARS-CoV-2 in mouse serum and plasma samples The assay uses electrochemiluminescence detection technology and provides quantitative results for IgA antibodies to the SARS-CoV-2 spike protein receptor binding domain and nucleocapsid protein This assay allows for high-throughput testing of potential vaccines and therapeutics in mouse models The kit includes reagents for analyzing 46 samples in duplicate along with calibrators and controls
Catalog No. 50-000-81164 Supplier Meso Scale Discovery Supplier No. K15572U2
Add to Cart
Add to Cart
Request a Quote
The Meso Scale Discovery V-PLEX SARS-CoV-2 Panel 2 Mouse IgA Kit is a multiplex immunoassay that measures IgA antibodies to SARS-CoV-2 in mouse serum and plasma samples The assay uses electrochemiluminescence detection technology and provides quantitative results for IgA antibodies to the SARS-CoV-2 spike protein receptor binding domain and nucleocapsid protein This assay allows for high-throughput testing of potential vaccines and therapeutics in mouse models The kit includes reagents for analyzing 46 samples in duplicate along with calibrators and controls