Learn More
Meso Scale Discovery VPLEX SARSCoV2 PNL 24 ACE2
SDP
Catalog No. 5000080242
Encompass
The Meso Scale Discovery SARS-CoV-2 Neutralization Antibody Assay Kit is a highly sensitive assay for detecting neutralizing antibodies against SARS-CoV-2 in serum and plasma samples It uses MSD electrochemiluminescence technology and provides quantitative measurement of SARS-CoV-2 neutralizing antibodies within 2 5 hours The assay has high specificity and sensitivity compared to traditional neutralization assays It includes a ready-to-use pre-coated plate and liquid stable reagents for convenience Overall this assay enables rapid high-throughput quantification of SARS-CoV-2 neutralizing antibodies
Catalog No. 50-000-80242 Supplier Meso Scale Discovery Supplier No. K15578U4
Add to Cart
Add to Cart
Request a Quote
The Meso Scale Discovery SARS-CoV-2 Neutralization Antibody Assay Kit is a highly sensitive assay for detecting neutralizing antibodies against SARS-CoV-2 in serum and plasma samples It uses MSD electrochemiluminescence technology and provides quantitative measurement of SARS-CoV-2 neutralizing antibodies within 2 5 hours The assay has high specificity and sensitivity compared to traditional neutralization assays It includes a ready-to-use pre-coated plate and liquid stable reagents for convenience Overall this assay enables rapid high-throughput quantification of SARS-CoV-2 neutralizing antibodies