Learn More
Meso Scale Discovery VPLEX SARSCoV2 PNL 24 IgA Kit
SDP
Catalog No. 5000078233
Encompass
The V-PLEX SARS-CoV-2 Panel 2 IgA Kit from Meso Scale Diagnostics detects IgA antibodies to SARS-CoV-2 in human serum and plasma using electrochemiluminescence on the QuickPlex SQ 120 instrument It includes a 96-well plate with SARS-CoV-2 antigens detection antibodies calibrators controls wash buffer read buffer and plate seals The assay takes about 3 hours and provides quantitative IgA results indicating recent or current infection aiding in understanding the immune response during COVID-19
Catalog No. 50-000-78233 Supplier Meso Scale Discovery Supplier No. K15577U2
Add to Cart
Add to Cart
Request a Quote
The V-PLEX SARS-CoV-2 Panel 2 IgA Kit from Meso Scale Diagnostics detects IgA antibodies to SARS-CoV-2 in human serum and plasma using electrochemiluminescence on the QuickPlex SQ 120 instrument It includes a 96-well plate with SARS-CoV-2 antigens detection antibodies calibrators controls wash buffer read buffer and plate seals The assay takes about 3 hours and provides quantitative IgA results indicating recent or current infection aiding in understanding the immune response during COVID-19