Learn More
Meso Scale Discovery VPLEX SARSCoV2 PNL 24 IgG Kit
SDP
Catalog No. 5000081415
Encompass
The Mesoscale K15575U-2 is a high-performance thermoelectric cooler for precision temperature control Measuring 40mm x 40mm x 3 4mm it offers a 15W heat pumping capacity at a 27 C difference This Peltier cooler has a no load voltage of 3 7V and can reach a 70 C temperature difference Made with Bismuth Telluride between ceramic substrates it needs a heat sink and fan for thermal management Its quiet compact design suits small instruments analytical devices laser systems and temperature-critical electronics
Catalog No. 50-000-81415 Supplier Meso Scale Discovery Supplier No. K15575U2
Add to Cart
Add to Cart
Request a Quote
The Mesoscale K15575U-2 is a high-performance thermoelectric cooler for precision temperature control Measuring 40mm x 40mm x 3 4mm it offers a 15W heat pumping capacity at a 27 C difference This Peltier cooler has a no load voltage of 3 7V and can reach a 70 C temperature difference Made with Bismuth Telluride between ceramic substrates it needs a heat sink and fan for thermal management Its quiet compact design suits small instruments analytical devices laser systems and temperature-critical electronics