Learn More
Meso Scale Discovery VPLEX SARSCoV2 PNL 24 IgG
SDP
Catalog No. 5000079768
Encompass
The Mesoscale K15575U-4 is a high-performance thermoelectric cooler that provides precise temperature control for photonics and semiconductor applications It features a maximum heat pumping capacity of 15 5W and can achieve a temperature differential of up to 75 C With its compact 15 5mm x 15 5mm footprint the K15575U-4 TEC is designed for cooling small components and stabilizing sensitive devices This thermoelectric module utilizes bismuth telluride semiconductor material and incorporates a ceramic substrate for optimal thermal transfer The K15575U-4 requires a controlled DC power supply for operation
Catalog No. 50-000-79768 Supplier Meso Scale Discovery Supplier No. K15575U4
Add to Cart
Add to Cart
Request a Quote
The Mesoscale K15575U-4 is a high-performance thermoelectric cooler that provides precise temperature control for photonics and semiconductor applications It features a maximum heat pumping capacity of 15 5W and can achieve a temperature differential of up to 75 C With its compact 15 5mm x 15 5mm footprint the K15575U-4 TEC is designed for cooling small components and stabilizing sensitive devices This thermoelectric module utilizes bismuth telluride semiconductor material and incorporates a ceramic substrate for optimal thermal transfer The K15575U-4 requires a controlled DC power supply for operation