Learn More
Meso Scale Discovery VPLEX SARSCoV2 PNL 24 MSE IgG
SDP
Catalog No. 5000081540
Encompass
The Mesoscale K15579U-2 is a high-performance thermoelectric cooler for photonics and semiconductor applications offering a 15W heat pumping capacity and a 0 C to 70 C range It combines a thermoelectric module fan and heat sink for efficient cooling in a compact 40 x 40 x 12 4mm design Operating on 12VDC it can be controlled via a thermistor or thermostat Its solid-state design ensures high reliability making it ideal for stabilizing laser diode temperatures and cooling detectors
Catalog No. 50-000-81540 Supplier Meso Scale Discovery Supplier No. K15579U2
Add to Cart
Add to Cart
Request a Quote
The Mesoscale K15579U-2 is a high-performance thermoelectric cooler for photonics and semiconductor applications offering a 15W heat pumping capacity and a 0 C to 70 C range It combines a thermoelectric module fan and heat sink for efficient cooling in a compact 40 x 40 x 12 4mm design Operating on 12VDC it can be controlled via a thermistor or thermostat Its solid-state design ensures high reliability making it ideal for stabilizing laser diode temperatures and cooling detectors