Learn More
Meso Scale Discovery VPLEX SARSCoV2 PNL 24 MSE IgM
SDP
Catalog No. 5000082334
Encompass
The Meso Scale Discovery SARS-CoV-2 Spike P-1 V-PLEX Kit is an electrochemiluminescence immunoassay for the quantitative detection of SARS-CoV-2 Spike P-1 protein in serum plasma and cell culture supernatant samples The assay uses MSD s proprietary V-PLEX technology and provides high sensitivity a broad dynamic range and rapid results in under 3 hours The kit includes a 96-well plate coated with capture antibodies detection antibodies conjugated to an electrochemiluminescent label calibrators controls and sample diluent The assay has high specificity for SARS-CoV-2 Spike P-1 with no observed cross-reactivity
Catalog No. 50-000-82334 Supplier Meso Scale Discovery Supplier No. K15580U4
Add to Cart
Add to Cart
Request a Quote
The Meso Scale Discovery SARS-CoV-2 Spike P-1 V-PLEX Kit is an electrochemiluminescence immunoassay for the quantitative detection of SARS-CoV-2 Spike P-1 protein in serum plasma and cell culture supernatant samples The assay uses MSD s proprietary V-PLEX technology and provides high sensitivity a broad dynamic range and rapid results in under 3 hours The kit includes a 96-well plate coated with capture antibodies detection antibodies conjugated to an electrochemiluminescent label calibrators controls and sample diluent The assay has high specificity for SARS-CoV-2 Spike P-1 with no observed cross-reactivity