Learn More
Meso Scale Discovery VPLEX SARSCoV2 PNL 25 ACE2
SDP
Catalog No. 5000082457
Encompass
The Mesoscale K15586U-4 is a 4-channel 16-bit 250 kS/s simultaneous sampling data acquisition module It has 4 analog voltage input channels with programmable input ranges of 10 V 5 V 2 V and 1 V The module has 16-bit resolution and a maximum sampling rate of 250 kS/s It can perform simultaneous sampling across all channels The K15586U-4 has built-in anti-aliasing filters and AC/DC coupling selection It is compatible with the Mesoscale RIO-47000 series chassis and streams data over PCIe to the host PC The module is suitable for multi-channel data logging and control applications
Catalog No. 50-000-82457 Supplier Meso Scale Discovery Supplier No. K15586U4
Add to Cart
Add to Cart
Request a Quote
The Mesoscale K15586U-4 is a 4-channel 16-bit 250 kS/s simultaneous sampling data acquisition module It has 4 analog voltage input channels with programmable input ranges of 10 V 5 V 2 V and 1 V The module has 16-bit resolution and a maximum sampling rate of 250 kS/s It can perform simultaneous sampling across all channels The K15586U-4 has built-in anti-aliasing filters and AC/DC coupling selection It is compatible with the Mesoscale RIO-47000 series chassis and streams data over PCIe to the host PC The module is suitable for multi-channel data logging and control applications