Learn More
Meso Scale Discovery VPLEX SARSCoV2 PNL 25 IgA Kit
SDP
Catalog No. 5000081541
Encompass
The V-PLEX SARS-CoV-2 Panel 2 IgA Kit is a serology test that detects IgA antibodies to SARS-CoV-2 in human serum and plasma It is an enzyme-linked immunosorbent assay (ELISA) that provides qualitative and semi-quantitative results The assay uses a recombinant SARS-CoV-2 antigen coated onto microwells to capture anti-SARS-CoV-2 IgA antibodies if present in the sample The kit includes reagents for 25 tests and can aid in identifying individuals with an adaptive immune response to SARS-CoV-2 indicating recent or prior infection
Catalog No. 50-000-81541 Supplier Meso Scale Discovery Supplier No. K15585U2
Add to Cart
Add to Cart
Request a Quote
The V-PLEX SARS-CoV-2 Panel 2 IgA Kit is a serology test that detects IgA antibodies to SARS-CoV-2 in human serum and plasma It is an enzyme-linked immunosorbent assay (ELISA) that provides qualitative and semi-quantitative results The assay uses a recombinant SARS-CoV-2 antigen coated onto microwells to capture anti-SARS-CoV-2 IgA antibodies if present in the sample The kit includes reagents for 25 tests and can aid in identifying individuals with an adaptive immune response to SARS-CoV-2 indicating recent or prior infection