Learn More
Meso Scale Discovery VPLEX SARSCoV2 PNL 25 IgA
SDP
Catalog No. 5000078287
Encompass
The Meso Scale Discovery V-PLEX SARS-CoV-2 Panel 2 IgA Kit is a serology assay that detects IgA antibodies to SARS-CoV-2 in human serum and plasma It uses electrochemiluminescence technology and provides quantitative results in under 3 hours The kit includes plates coated with SARS-CoV-2 antigens detection antibodies calibrators controls wash buffers readout buffer and plate seals It has high sensitivity and specificity for detecting recent or prior infection with SARS-CoV-2 This assay aids in understanding immune response and tracking infection rates
Catalog No. 50-000-78287 Supplier Meso Scale Discovery Supplier No. K15585U4
Add to Cart
Add to Cart
Request a Quote
The Meso Scale Discovery V-PLEX SARS-CoV-2 Panel 2 IgA Kit is a serology assay that detects IgA antibodies to SARS-CoV-2 in human serum and plasma It uses electrochemiluminescence technology and provides quantitative results in under 3 hours The kit includes plates coated with SARS-CoV-2 antigens detection antibodies calibrators controls wash buffers readout buffer and plate seals It has high sensitivity and specificity for detecting recent or prior infection with SARS-CoV-2 This assay aids in understanding immune response and tracking infection rates