Learn More
Meso Scale Discovery VPLEX SARSCoV2 PNL 25 IgG Kit
SDP
Catalog No. 5000080251
Encompass
The Meso Scale Discovery K15583U-2 is a 96-well plate for measuring phospho-STAT3 (Tyr705) in cell lysates It uses electrochemiluminescence technology for high sensitivity and broad dynamic range The plate is pre-coated with capture antibody and comes with detection antibody and read buffer It allows quantification of STAT3 phosphorylation and monitoring of STAT3 pathway activation The kit includes 1 plate detection antibody read buffer sample diluent and plate seals It is suitable for cell signaling research and drug discovery applications
Catalog No. 50-000-80251 Supplier Meso Scale Discovery Supplier No. K15583U2
Add to Cart
Add to Cart
Request a Quote
The Meso Scale Discovery K15583U-2 is a 96-well plate for measuring phospho-STAT3 (Tyr705) in cell lysates It uses electrochemiluminescence technology for high sensitivity and broad dynamic range The plate is pre-coated with capture antibody and comes with detection antibody and read buffer It allows quantification of STAT3 phosphorylation and monitoring of STAT3 pathway activation The kit includes 1 plate detection antibody read buffer sample diluent and plate seals It is suitable for cell signaling research and drug discovery applications