Learn More
Meso Scale Discovery VPLEX SARSCoV2 PNL 25 IgG
SDP
Catalog No. 5000081419
Encompass
The Meso Scale Discovery K15583U-4 is a 96-well plate coated with a proprietary carbon electrode sensor that enables label-free detection of proteins and other biomolecules using electrochemiluminescence (ECL) It is designed for high-sensitivity quantitative analysis and multiplexing of up to 10 protein biomarkers from cell lysates and biofluids like serum plasma and CSF The carbon electrode surface has high protein binding capacity and low non-specific binding It is compatible with commonly used ECL reagents and detection instruments This product enables rapid and accurate protein biomarker profiling for life science research and drug development applications
Catalog No. 50-000-81419 Supplier Meso Scale Discovery Supplier No. K15583U4
Add to Cart
Add to Cart
Request a Quote
The Meso Scale Discovery K15583U-4 is a 96-well plate coated with a proprietary carbon electrode sensor that enables label-free detection of proteins and other biomolecules using electrochemiluminescence (ECL) It is designed for high-sensitivity quantitative analysis and multiplexing of up to 10 protein biomarkers from cell lysates and biofluids like serum plasma and CSF The carbon electrode surface has high protein binding capacity and low non-specific binding It is compatible with commonly used ECL reagents and detection instruments This product enables rapid and accurate protein biomarker profiling for life science research and drug development applications